missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Methylnicotinate, TRC

Product Code. 30469861
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469861 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469861 Supplier Toronto Research Chemicals Supplier No. TRCM3230051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 5-Methylnicotinate
CAS 29681-45-6
Molecular Formula C8 H9 N O2
Quantity 1 g
Synonym 3-Pyridinecarboxylic acid','5-methyl-','methyl ester, Nicotinic acid','5-methyl-','methyl ester (8CI), 5-Methylnicotinic acid methyl ester, 5-Methylpyridine-3-carboxylic acid methyl ester, Methyl 5-methyl-3-pyridinecarboxylate, 5-Methyl-3-pyridinecarboxylic acid methyl ester, Methyl 5-methylpyridine-3-carboxylate, Methyl 5-methylnicotinate, NSC 83076
InChI Formula InChI=1S/C8H9NO2/c1-6-3-7(5-9-4-6)8(10)11-2/h3-5H,1-2H3
SMILES COC(=O)c1cncc(C)c1
IUPAC Name methyl 5-methylpyridine-3-carboxylate
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.