Learn More
Methyl 9-Hydroxynon-4-enoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 9-Hydroxynon-4-enoate |
| CAS | 87238-54-8 |
| Molecular Formula | C10H18O3 |
| Quantity | 2.5 mg |
| Synonym | (E)-9-Hydroxy-4-nonenoic Acid Methyl Ester |
| InChI Formula | InChI=1S/C10H18O3/c1-13-10(12)8-6-4-2-3-5-7-9-11/h2,4,11H,3,5-9H2,1H3/b4-2+ |
| SMILES | COC(=O)CC\C=C\CCCCO |
| IUPAC Name | methyl (E)-9-hydroxynon-4-enoate |
| Molecular Weight (g/mol) | 186.25 |
| Formula Weight | 186.1256 |