missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl alpha-Hydroxy-6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-acetate, Mikromol™
Description
Methyl alpha-Hydroxy-6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-acetate
Specifications
Specifications
| Chemical Name or Material | Methyl alpha-Hydroxy-6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-acetate |
| CAS | 258273-49-3 |
| Analyte or Component Names | Methyl alpha-Hydroxy-6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl alpha-Hydroxy-6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine-3-acetate |
| InChI Formula | InChI=1S/C18H18N2O3/c1-11-4-7-13(8-5-11)15-16(17(21)18(22)23-3)20-10-12(2)6-9-14(20)19-15/h4-10,17,21H,1-3H3 |
| SMILES | COC(=O)C(O)c1c(nc2ccc(C)cn12)c3ccc(C)cc3 |
| IUPAC Name | methyl 2-hydroxy-2-[6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?