missing translation for 'onlineSavingsMsg'
Learn More

Methyl β-D-thiogalactoside, 98%, Thermo Scientific™

Catalog Number p-7077799
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
155-30-6
C7H14O5S
210.25
LZFNFLTVAMOOPJ-PZRMXXKTSA-N
methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose
10036019
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol
CSC1C(C(C(C(O1)CO)O)O)O
This item is not returnable. View return policy
155-30-6
C7H14O5S
210.25
LZFNFLTVAMOOPJ-PZRMXXKTSA-N
methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose
10036019
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol
CSC1C(C(C(C(O1)CO)O)O)O
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: soluble.
Formula Weight 210.25
Percent Purity 98%
Chemical Name or Material Methyl β-D-thiogalactoside
compliance-icons compliance-icons
Product Identifier
  • Methyl beta-D-thiogalactoside
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'