missing translation for 'onlineSavingsMsg'
Learn More

Methyl-d3-triphenylphosphonium Bromide, CDN

Product Code. 30313804
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313804 10 g 10g
30309187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30313804 Supplier CDN Supplier No. CDND084310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl-d3-triphenylphosphonium Bromide
CAS 1787-44-6
Molecular Formula C19 D3 H15 P . Br
Quantity 10 g
Synonym Phosphonium, methyl-d3-triphenyl-, bromide (1:1) (ACI), Methyl-d3-triphenylphosphonium bromide (7 CI), Phosphonium, methyl-d3-triphenyl-, bromide (8 CI, 9 CI), Trideuteriomethyltriphenylphosphonium bromide, (Trideuteromethyl)triphenylphosphonium bromide, Triphenyl(trideuteriomethyl)phosphonium bromide, (Methyl)triphenylphosphonium bromide-d3, Methyltriphenylphosphonium-d3 bromide, Methyltriphenylphosphonium-D3 (methyl-D3) bromide
InChI Formula InChI=1 S/C19H18P.BrH/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16 H,1H3;1 H/q+1;/p-1/i1D3;
SMILES [Br-].[2 H]C([2 H])([2 H])[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name triphenyl(trideuteriomethyl)phosphanium;bromide
Molecular Weight (g/mol) 360.243
Formula Weight 359.052 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.