Learn More
Methyl DL-Mandelate, TRC
Specifications
Specifications
| CAS | 4358-87-6 |
| Molecular Formula | C9 H10 O3 |
| Quantity | 1 g |
| Synonym | Benzeneacetic acid, α-hydroxy-, methyl ester, Benzeneacetic acid, α-hydroxy-, methyl ester, (±)-, Mandelic acid, methyl ester, DL- (8CI), (±)-Mandelic acid methyl ester, (±)-Methyl mandelate, 2-Hydroxy-2-phenylethanoic acid methyl ester, Methyl (RS)-α-hydroxybenzeneacetate, Methyl (hydroxy)(phenyl)acetate, Methyl (±)-α-hydroxyphenylacetate, Methyl 2-hydroxy-2-phenylacetate, Methyl 2-hydroxy-2-phenylethanoate, Methyl DL-mandelate, Methyl mandelate, Methyl phenylglycolate, Methyl α-hydroxyphenylacetate, NSC 6539, α-Hydroxybenzeneacetic acid methyl ester |
| InChI Formula | InChI=1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3 |
| SMILES | COC(=O)C(O)c1ccccc1 |
| IUPAC Name | methyl 2-hydroxy-2-phenylacetate |
| Molecular Weight (g/mol) | 166.17 |
| Formula Weight | 166.063 |
| Chemical Name or Material | Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate) |