missing translation for 'onlineSavingsMsg'
Learn More

Methyl Hexanoate, TRC

Product Code. 30135748
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135748 500 g 500g
30007532 250 g 250g
30005279 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135748 Supplier Toronto Research Chemicals Supplier No. TRCM312065500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-70-7
Molecular Formula C7 H14 O2
Molecular Weight (g/mol) 130.18
Synonym Hexanoic Acid Methyl Ester, Hexanoic acid, methyl ester, Agnique 610G, Caproic acid methyl ester, Methyl caproate, Methyl capronate, Methyl hexanoate, Methyl hexoate, Methyl n-hexanoate, NSC 5023, Pastell M 6
IUPAC Name methyl hexanoate
SMILES CCCCCC(=O)OC

Specifications

Quantity 500 g
InChI Formula InChI=1S/C7H14O2/c1-3-4-5-6-7(8)9-2/h3-6H2,1-2H3
Formula Weight 130.0994
Chemical Name or Material Caproic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.