Learn More
Methyl Indole-3-acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Indole-3-acetate |
| CAS | 1912-33-0 |
| Molecular Formula | C11 H11 N O2 |
| Quantity | 5 g |
| Synonym | 1H-Indole-3-acetic acid','methyl ester (9CI','ACI), Indole-3-acetic acid','methyl ester (6CI','7CI','8CI), (1H-Indol-3-yl)acetic acid methyl ester, IAA methyl ester, Indole-3-methyl acetate, Indolyl-3-acetic acid methyl ester, Methyl 1H-indol-3-ylacetate, Methyl 1H-indole-3-acetate, Methyl 2-(1H-indol-3-yl)acetate, Methyl indol-3-ylacetate, Methyl indole-3-acetate, Methyl β-indoleacetate, NSC 63806, β-Indolylacetic acid methyl ester, 1H-Indole-3-acetic Acid Methyl Ester |
| InChI Formula | InChI=1S/C11H11NO2/c1-14-11(13)6-8-7-12-10-5-3-2-4-9(8)10/h2-5,7,12H,6H2,1H3 |
| SMILES | COC(=O)Cc1c[nH]c2ccccc12 |
| IUPAC Name | methyl 2-(1H-indol-3-yl)acetate |
| Molecular Weight (g/mol) | 189.21 |
| Formula Weight | 189.079 |