Learn More
Methyl isovalerate, 98%
Product Code. 11332436
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332436 250 g 250g
11322436 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11332436 Supplier Thermo Scientific Alfa Aesar Supplier No. L06241.30
Request Bulk or Custom Format
CAS: 556-24-1 | C6H12O2 | 116.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC
Specifications
Density 0.88
Boiling Point 115°C to 117°C
Flash Point 16°C (60°F)
Packaging Glass bottle
Refractive Index 1.3936
Quantity 250 g
UN Number UN2400
Beilstein 1699922
Specific Gravity 0.88
Formula Weight 116.17
Percent Purity 98%
Chemical Name or Material Methyl isovalerate