missing translation for 'onlineSavingsMsg'
Learn More

Methyl Octahydro-1H-indole-2-carboxylate Hydrochloride, TRC

Product Code. 30336387
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336387 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336387 Supplier Toronto Research Chemicals Supplier No. TRCB499388250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Octahydro-1H-indole-2-carboxylate Hydrochloride
CAS 195878-03-6
Molecular Formula C10H17NO2 . HCl
Quantity 250 mg
Synonym 1H-Indole-2-carboxylic acid, octahydro-, methyl ester, hydrochloride (1:1) (ACI), 1H-Indole-2-carboxylic acid, octahydro-, methyl ester, hydrochloride (9CI), Methyl octahydro-1H-indole-2-carboxylate hydrochloride, Octahydro-1H-indole-2-carboxylic acid methyl ester hydrochloride
InChI Formula InChI=1S/C10H17NO2.ClH/c1-13-10(12)9-6-7-4-2-3-5-8(7)11-9;/h7-9,11H,2-6H2,1H3;1H
SMILES Cl.COC(=O)C1CC2CCCCC2N1
IUPAC Name methyl 2,3,3a,4,5,6,7,7a-octahydro-1H-indole-2-carboxylate;hydrochloride
Molecular Weight (g/mol) 219.71
Formula Weight 219.1026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.