Learn More
Methyl palmitate, 99%, analytical standard for GC
Product Code. p-7077837 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15520644
Brand: Thermo Scientific Acros 460390010
Request Bulk or Custom Format
Edge
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC
Specifications
Melting Point 28.0°C
Boiling Point 338.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)14CO2CH3
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: solubel in alcohol, ether
Formula Weight 270.45
Percent Purity 99%
Chemical Name or Material Methyl palmitate