missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-Mandelate, TRC

Product Code. 30481984
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481984 25 g 25g
30472200 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481984 Supplier Toronto Research Chemicals Supplier No. TRCM31745525G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20698-91-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.17
Synonym (alphaR)-alpha-Hydroxybenzeneacetic Acid Methyl Ester, (R)-(-)-Mandelic Acid Methyl Ester, (-)-Mandelic Acid Methyl Ester, (-)-Methyl Mandelate, (R)-(-)-Mandelic Acid Methyl Ester, (R)-(-)-Methyl 2-Hydroxy-2-phenylacetate, (R)-Mandelic Acid Methyl Ester, (R)-Methyl (Hydroxy)(phenyl)acetate, (R)-Methyl Mandelate, (R)-alpha-Hydroxybenzeneacetic Acid Methyl Ester, (alphaR)-alpha-Hydroxybenzeneacetic Acid Methyl Ester, D-(-)-Mandelic Acid Methyl Ester, D-(-)-Methyl Mandelate, Methyl (R)-2-Hydroxy-2-phenylacetate, Methyl (R)-Mandelate, Methyl (R)-alpha-Hydroxybenzeneacetate, Methyl (R)-alpha-Phenylglycolate, Methyl D-Mandelate, Methyl l-Mandelate
IUPAC Name methyl (2R)-2-hydroxy-2-phenylacetate
SMILES COC(=O)[C@H](O)c1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3/t8-/m1/s1
Formula Weight 166.063
Chemical Name or Material Methyl (R)-(-)-Mandelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.