missing translation for 'onlineSavingsMsg'
Learn More

Methyl tert-butyl ether (MTBE) 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30325887
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325887 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325887 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010523ME
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl tert-butyl ether
CAS 1634-04-4
Analyte or Component Names Methyl tert-butyl ether
Shipping Condition Room Temperature
Molecular Formula C5 H12 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Propane, 2-methoxy-2-methyl-, Ether, tert-butyl methyl (6CI,7CI,8CI), 2-Methoxy-2-methylpropane, 1,1-Dimethylethyl methyl ether, 2-Methyl-2-methoxypropane, Driveron, MTBE, Methyl 1,1-dimethylethyl ether, Methyl tert-butyl ether, Methyl tertiary butyl ether, t-Butyl methyl ether, tert-Butoxymethane, tert-Butyl methyl ether
InChI Formula InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3
SMILES COC(C)(C)C
IUPAC Name 2-methoxy-2-methylpropane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 88.15
Quantity 1 mL
Formula Weight 88.0888 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.