missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-hexenoate, 97%

Product Code. 11357955
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357955 50 g 50g
11347955 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357955 Supplier Thermo Scientific Alfa Aesar Supplier No. L08030.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13894-63-8 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13894-63-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00048798
InChI Key GFUGBRNILVVWIE-AATRIKPKSA-N
Synonym methyl 2-hexenoate, methyl e-2-hexenoate, methyl trans-2-hexenoate, methyl e-hex-2-enoate, methyl 2e-hexenoate, methyl 2e-hex-2-enoate, 2-hexenoic acid, methyl ester, e, methyl 2e-2-hexenoate, 2e-2-hexenoic acid methyl ester, unii-x2ddu82182
PubChem CID 5364409
IUPAC Name methyl (E)-hex-2-enoate
SMILES CCCC=CC(=O)OC

Specifications

Density 0.915
Boiling Point 57°C to 58°C (13 mmHg)
Flash Point 45°C (113°F)
Odor Fruit-like
Refractive Index 1.4345
Quantity 50 g
UN Number UN3272
Beilstein 1700768
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Methyl trans-2-hexenoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.