Learn More
Methyl2-(4-methylphenylsulfonamido)acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl2-(4-methylphenylsulfonamido)acetate |
| CAS | 2645-02-5 |
| Molecular Formula | C10H13NO4S |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C10H13NO4S/c1-8-3-5-9(6-4-8)16(13,14)11-7-10(12)15-2/h3-6,11H,7H2,1-2H3 |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)OC |
| IUPAC Name | methyl 2-[(4-methylphenyl)sulfonylamino]acetate |
| Molecular Weight (g/mol) | 243.28 |
| Formula Weight | 243.0565 |