missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-4813557
127.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-87-2
C7H14
98.19
MFCD00001497
UAEPNZWRGJTJPN-UHFFFAOYSA-N
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
7962
methylcyclohexane
CC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Melting Point -126.3°C
Density 0.7700g/mL
Boiling Point 101.0°C
Flash Point −3°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CH3
Refractive Index 1.4210 to 1.4230
Quantity 1 L
Merck Index 15,6118
Specific Gravity 0.77
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
Formula Weight 98.19
Percent Purity 99%
Grade Spectroscopy
Chemical Name or Material Methylcyclohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'