missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromopyridine-2-carboxylate, 97%, ACROS Organics™

Brand:  Acros Organics 436870050

Additional Details : CAS : 26218-75-7 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11906361

  • € 115.00 / 5g


Estimated Shipment: 05-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

26218-75-7
216.03
SGNCOKUHMXLGAH-UHFFFAOYSA-N
16217886
COC(=O)C1=NC(Br)=CC=C1
C7H6BrNO2
MFCD06203934
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate
methyl 6-bromopyridine-2-carboxylate

Specifications

Methyl 6-bromopyridine-2-carboxylate
Glass bottle
White to Yellow
5g
96% min. (HPLC)
MFCD06203934
SGNCOKUHMXLGAH-UHFFFAOYSA-N
methyl 6-bromopyridine-2-carboxylate
16217886
Crystalline Powder or Needles
Authentic
1% Max.
94.6°C to 95°C
26218-75-7
C7H6BrNO2
methyl 6-bromopicolinate, 6-bromopyridine-2-carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, methyl 6-bromo-2-pyridinecarboxylate, 6-bromo-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 2-bromo-6-methoxycarbonyl pyridine, methyl 2-bromo-6-nicotinate, methyl-6-bromopyridine-2-carboxylate, methyl-6-bromo-2-pyridine carboxylate
COC(=O)C1=NC(Br)=CC=C1
216.03
216.03
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl 6-bromopyridine-2-carboxylate, 97%

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates