missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylglyoxal Antibody (9E7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59368UV
This item is not returnable.
View return policy
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Methylglyoxal | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLO1 | |
| Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μL | |
| Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 90000000 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 2-oxopropanal, Pyruvaldehyde | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human, Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction