missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylglyoxal Antibody (9E7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 188.00 - € 641.55
Specifications
| Antigen | Methylglyoxal |
|---|---|
| Clone | 90000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18061376
|
Novus Biologicals
NBP2-59368 |
100 μg |
€ 679.00 € 641.55 / 100µg Save € 37.45 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645868
|
Novus Biologicals
NBP2-59368-25ug |
25 μg |
€ 188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Methylglyoxal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 2739 | |
| Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. |
| 90000000 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 2-oxopropanal, Pyruvaldehyde | |
| GLO1 | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title