Learn More
(Methylthio)acetic acid, 98%
Product Code. 11420737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11420737 1 g 1g
11430737 5 g 5g
11440737 25 g 25g
3 options
Product Code. 11420737
Brand: Thermo Scientific Alfa Aesar B22531.03
Request Bulk or Custom Format
CAS: 2444-37-3 | C3H6O2S | 106.139 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2444-37-3
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00075444
InChI Key HGTBAIVLETUVCG-UHFFFAOYSA-N
Synonym methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
PubChem CID 75551
ChEBI CHEBI:47870
IUPAC Name 2-methylsulfanylacetic acid
SMILES CSCC(=O)O
Specifications
Melting Point 13°C to 14°C
Density 1.224
Boiling Point 99°C to 100°C (4 mmHg)
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.495
Quantity 1 g
Beilstein 1740201
Formula Weight 106.15
Percent Purity 98%
Chemical Name or Material (Methylthio)acetic acid