missing translation for 'onlineSavingsMsg'
Learn More

Methyltitanium(IV) triisopropoxide, 1M solution in THF

Product Code. 10360424 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10360424 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10360424 Supplier Thermo Scientific Acros Supplier No. 384610500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18006-13-8 | C10H24O3Ti | 240.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 18006-13-8, 109-99-9
CAS Min % 26.84, 70.48
CAS Max % 29.52, 73.16
Density 0.8950g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging Glass Bottle
Molecular Formula C10H24O3Ti
MDL Number MFCD03411233
Quantity 50 mL
Specific Gravity 0.895
Synonym titanium, methyltripropoxy, titanium,methyltris 2-propanolato-, t-4, methyltitanium triisopropoxide, 1m in tetrahydrofuran
InChI Key QBLPAZNBFFURKB-UHFFFAOYSA-N
SMILES [CH3-].CC(C)O.CC(C)O.CC(C)O.[Ti]
IUPAC Name carbanide;propan-2-ol;titanium
Molecular Weight (g/mol) 240.18
PubChem CID 144064
Formula Weight 240.18
Chemical Name or Material Methyltitanium(IV) triisopropoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyltitanium(IV) triisopropoxide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.