Learn More
Metrizoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Metrizoic Acid |
| CAS | 1949-45-7 |
| Molecular Formula | C12H11I3N2O4 |
| Quantity | 2 g |
| Synonym | 3-(Acetylamino)-5-(acetylmethylamino)-2,4,6-triiodobenzoic Acid, 3-Acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic Acid, 3,5-Diacetamido-N-methyl-2,4,6-triiodobenzoic Acid, Metrizoate |
| InChI Formula | InChI=1S/C12H11I3N2O4/c1-4(18)16-10-7(13)6(12(20)21)8(14)11(9(10)15)17(3)5(2)19/h1-3H3,(H,16,18)(H,20,21) |
| SMILES | CN(C(=O)C)c1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I |
| IUPAC Name | 3-acetamido-5-[acetyl(methyl)amino]-2,4,6-triiodobenzoic acid |
| Molecular Weight (g/mol) | 627.94 |
| Formula Weight | 627.7853 |