Learn More
Metro™ Double-Deep qwikTRAK™ Stationary End Unit Kits
Connects end units to tracks and anchors tracks to floor
Brand:  Metro™ LBTES
Additional Details : Weight : 26.85600kg
 View more versions of this product
Product Code. 15936981
  • € 2835.00 / Each
Estimated Shipment: 09-05-2023
to see stock.
Add to basket
Description
Description
  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
  • Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Specifications
Stationary end unit kit
33.5 kg
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor
Stainless Steel
73.6 lb.
Metro™ Super Erecta™, Super Adjustable™ shelving system