Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
Product Code. 15967281
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
None
Unit Size:
Each
Product Code. 15967281
Brand: Metro™ TTA21S
Only null left
Add to basket
Add to basket
Includes
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
High-density, easy access storage
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts
Order Info
Shelves are ordered separately
Specifications
Product Type Stationary Intermediate Unit Kit
Finish None
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Height (English) 86 in.
Height (Metric) 2185 mm
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 8 lb.
Weight (Metric) 3.6 kg
Width (English) Shelf 21 in.
Width (Metric) Shelf 53 cm
Material Stainless Steel