missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ METT10D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29527
This item is not returnable.
View return policy
Description
METT10D Polyclonal antibody specifically detects METT10D in Human,Mouse samples. It is validated for Western Blot
Specifications
| METT10D | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79066 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D | |
| Between 375 and 425 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction