missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METT10D Antibody (OTI3B5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72654CL1
This item is not returnable.
View return policy
Description
METT10D Monoclonal antibody specifically detects METT10D in Human,Monkey,Mouse,Primate samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| METT10D | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D | |
| Full length human recombinant protein of human METT10D(NP_076991) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey, Mouse, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI3B5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79066 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction