missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Antibody (9C2F2), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06651AF594
This item is not returnable.
View return policy
Description
METTL1 Monoclonal antibody specifically detects METTL1 in Human samples. It is validated for ELISA, ELISA, ELISA
Specifications
| METTL1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 9C2F2 | |
| ELISA, Sandwich ELISA, Sandwich ELISA Detection | |
| C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human METTL1 (NP_005362.3; Asp 32-Gln 265). | |
| 0.1 mL | |
| Signal Transduction | |
| 4234 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction