missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL10 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21446CL1
This item is not returnable.
View return policy
Description
METTL10 Polyclonal antibody specifically detects METTL10 in Human samples. It is validated for Immunoprecipitation,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| METTL10 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 399818 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| C10orf138, chromosome 10 open reading frame 138, EC 2.1.1.-, Em:AC068896.3, FLJ13019, methyltransferase like 10, methyltransferase-like protein 10 | |
| Produced in rabbits immunized with E. coli-derived Human METTL10 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction