missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL14 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18489H
This item is not returnable.
View return policy
Description
METTL14 Polyclonal antibody specifically detects METTL14 in Human samples. It is validated for Western Blot
Specifications
| METTL14 | |
| Polyclonal | |
| Western Blot | |
| EC 2.1.1, EC 2.1.1.-, KIAA1627methyltransferase-like protein 14, methyltransferase like 14 | |
| Recognizes region between residues Between between 25 and 75.. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 57721 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction