missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543284
This item is not returnable.
View return policy
Description
Peptide sequence: MLRYPYFCRM YKECLSCWLE SGIPNLGVWP NRIHTTAEKY REYEAREQTD Sequence homology: Human: 100%.
METT5D1 belongs to the methyltransferase superfamily, mraW family. METT5D1 is a probable S-adenosyl-L-methionine-dependent methyltransferase.
Specifications
| METTL15 | |
| Polyclonal | |
| Unconjugated | |
| METTL15 | |
| methyltransferase 5 domain containing 1; methyltransferase 5 domain-containing protein 1; methyltransferase like 15; METT5D1; METTL15; Probable methyltransferase-like protein 15; probable S-adenosyl-L-methionine-dependent methyltransferase METT5D1; putative S-adenosyl-L-methionine-dependent methyltransferase METT5D1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 196074 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6NJ78 | |
| METTL15 | |
| Synthetic peptide directed towards the N-terminal of human METT5D1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction