Learn More
Invitrogen™ METTL21B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139805
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GILAALQGGD VTITDLPLAL EQIQGNVQAN VPAGGQAQVR ALSWGIDHHV FPANYDLVLG ADIVYLEPT Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.
Specifications
| METTL21B | |
| Polyclonal | |
| Unconjugated | |
| EEF1AKMT3 | |
| EEF1A lysine methyltransferase 3; EEF1AKMT3; FAM119B; family with sequence similarity 119, member B; HCA557A; Hepatocellular carcinoma-associated antigen 557a; hepatocellularcarcinoma-associated antigen HCA557a; methyltransferase like 21B; methyltransferase-like protein 21B; METTL21B; Protein-lysine methyltransferase METTL21B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96AZ1 | |
| EEF1AKMT3 | |
| Recombinant protein corresponding to Human EEF1AKMT3. Recombinant protein control fragment (Product #RP-108855). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.