missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563787
This item is not returnable.
View return policy
Description
Immunogen sequence: DEDGDLDVVR RPRAASDSNP AGPLRDKVHP MILAQEEDDV LGEEAQGSPH DIIRIEHTMA TPLEDVGKQV Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Protein N-lysine methyltransferase. In vitro methylates KIN.
Specifications
| METTL22 | |
| Polyclonal | |
| Unconjugated | |
| METTL22 | |
| C16orf68; C25H16orf68; LP8272; methyltransferase like 22; methyltransferase-like protein 22; Mettl22; RGD1311433 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUU2 | |
| METTL22 | |
| Recombinant protein corresponding to Human METTL22. Recombinant protein control fragment (Product #RP-103360). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction