missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL3 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | METTL3 |
|---|---|
| Clone | 4B4A3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235941
|
Proteintech
67733-2-IG-150UL |
150 μL |
€ 361.81
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235485
|
Proteintech
67733-2-IG-20UL |
20 μL |
€ 133.74
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The METTL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human METTL3. This antibody recognizes human, mouse antigen. The METTL3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| METTL3 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 56339 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 4B4A3 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| IME4; M6A; MGC4336; MT-A70; Spo8 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title