missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569560
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%; Zebrafish: 93%.
METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
Specifications
| METTL4 | |
| Polyclonal | |
| Unconjugated | |
| METTL4 | |
| 2410198H06Rik; A730091E08Rik; AV296509; HsT661; methyltransferase like 4; methyltransferase-like protein 4; METTL4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; methyltransferase-like protein 4; RGD1306451 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 316731 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| METTL4 | |
| synthetic peptide directed towards the C terminal region of human METTL4. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction