Learn More
METTL7A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17092-1-AP-20UL
 View more versions of this product
Product Code. 16856934
  • € 145.00 / 20µL
Description
The METTL7A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human METTL7A. This antibody recognizes human, mouse, rat antigen. The METTL7A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
METTL7A
Polyclonal
Unconjugated
20 μL
25840
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse