Learn More
ING5 Antibody (06), Alexa Fluor™ 647 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06215AF647
Product Code. 18561622
  • € 423.00 / 0.10mL
Description
ING5 Monoclonal antibody specifically detects ING5 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ING5
Monoclonal
Alexa Fluor 647
inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ING5 (Q8WYH8-2; Glu36-Gln84).
0.1 mL
Cancer, Tumor Suppressors
84289
Store at 4°C in the dark.
IgG
ELISA
06
50mM Sodium Borate
Mouse
Protein A purified