missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Detachable Cables
Accessories for use with electrodes.
POA
Specifications
Color | Gray |
---|---|
Connector Types | SC-LEMO |
For Use With (Equipment) | Amperometric Titration and Double Pin Platinum Electrode |
Type | Cable |
Description
For use with electrodes.Specifications
Gray | |
SC-LEMO | |
Amperometric Titration and Double Pin Platinum Electrode | |
Cable |