Learn More
METTLER TOLEDO™ InLab™ Versatile pH Electrode Replaceable Ceramic Junctions
Shop All Mettler Toledo Products
Click to view available options
Junction:
Ceramic
Unit Size:
Pack of 10
Description
- Accessory
- For use with InLab™ Versatile and InLab™ Versatile Pro
Specifications
Specifications
| Junction | Ceramic |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | Set of replaceable junctions |
| For Use With (Application) | Alternative for measurements in simple samples, offer an outstanding price/performance ratio |
| For Use With (Equipment) | InLab designations versatile and versatile Pro pH electrode |
| Includes | 10 Ceramic junction |
Product Content Correction
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido