Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
Product Code. 16309835 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
Beaker
Drying Cup
Septa
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
16309835 Drying Cup Each
15807641 Septa Pack of 10
12750914 Beaker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16309835 Supplier Mettler Toledo™ Supplier No. 00023961
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with V20 Karl Fischer Autotitrators.
  • Accessory for DV1000 titration burettes
  • For titrant protection from moisture or CO2
  • With cover
TRUSTED_SUSTAINABILITY
Specifications
Type Drying Cup
For Use With (Application) Water content determination
Weight (Metric) 453.5 g