Learn More
Mettler Toledo™ Temperature Calibration Kits
Product Code. 10588754 Shop All Mettler Toledo Products
Change view
Click to view available options
Without Certificate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10588754 Without Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10588754 Supplier Mettler Toledo™ Supplier No. 214455
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
  • Adjustment disc with precision thermometer
  • Sample pan handler
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Halogen Moisture Analyzer
Includes Adjustment disc with precision thermometer, sample
Type Temperature Calibration Kit