Learn More
Mevastatin Acid Sodium Salt, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Mevastatin Acid Sodium Salt
Specifications
Specifications
| Chemical Name or Material | Mevastatin Acid Sodium Salt |
| CAS | 99782-89-5 |
| Analyte or Component Names | Mevastatin Acid Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H35 O6 . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | (βR,ΔR,1S,2S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,Δ-dihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic Acid Sodium Salt (1:1), 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ-dihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-, monosodium salt, [1S-[1α(βS*,ΔS*),2α,8β(R*),8aα]]-, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ-dihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-, monosodium salt, (βR,ΔR,1S,2S,8S,8aR)- (9CI), ML 236B sodium salt, Mevastatin Acid Sodium Salt |
| InChI Formula | InChI=1S/C23H36O6.Na/c1-4-14(2)23(28)29-20-7-5-6-16-9-8-15(3)19(22(16)20)11-10-17(24)12-18(25)13-21(26)27;/h6,8-9,14-15,17-20,22,24-25H,4-5,7,10-13H2,1-3H3,(H,26,27);/q;+1/p-1/t14-,15-,17+,18+,19-,20-,22-;/m0./s1 |
| SMILES | [Na+].CC[C@H](C)C(=O)O[C@H]1CCC=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| IUPAC Name | sodium;(3R,5R)-7-[(1S,2S,8S,8aR)-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Show More |