missing translation for 'onlineSavingsMsg'
Learn More

Mevastatin Hydroxy Acid t-Butylamine Salt, TRC

Product Code. 30469277
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469277 Supplier Toronto Research Chemicals Supplier No. TRCM34051550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mevastatin Hydroxy Acid t-Butylamine Salt
CAS 262285-80-3
Molecular Formula C27H47NO6
Quantity 50 mg
Synonym (betaR,deltaR,1S,2S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-beta,delta-dihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoate 2-Methyl-2-propanamine
InChI Formula InChI=1S/C23H36O6.C4H11N/c1-4-14(2)23(28)29-20-7-5-6-16-9-8-15(3)19(22(16)20)11-10-17(24)12-18(25)13-21(26)27;1-4(2,3)5/h6,8-9,14-15,17-20,22,24-25H,4-5,7,10-13H2,1-3H3,(H,26,27);5H2,1-3H3/t14-,15-,17+,18+,19-,20-,22-;/m0./s1
SMILES CC[C@H](C)C(=O)O[C@H]1CCC=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)O)[C@@H]12.CC(C)(C)N
IUPAC Name (3R,5R)-7-[(1S,2S,8S,8aR)-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic acid;2-methylpropan-2-amine
Molecular Weight (g/mol) 481.67
Formula Weight 481.3403
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.