Learn More
VITLAB™ MF Instrument Tray
Click to view available options
Dimensions (L x W x H):
240 x 180 x 17 mm
Unit Size:
Pack of 5
Description
- White
- Flat shape
- Rounded corners
- Smooth surfaces
- Easy to clean
- Self-supporting
Specifications
Specifications
| Dimensions (L x W x H) | 240 x 180 x 17 mm |
| Color | White |
| Height (Metric) | 17 mm |
| Length (Metric) | 240 mm |
| Material | Melamine Formaldehyde |
| Type | Instrument Tray |
| Width (Metric) | 180 mm |
| For Use With | Practical tray for instruments, tools, and sensitive utensils |
| No. per Pack | 5/Pk. |