missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559958
This item is not returnable.
View return policy
Description
Immunogen sequence: KELEENKRSL AALDALNTDD ENDEEEYEAW KVRELKRIKR DREDREALEK EKAEIERMRN LTEEERRAEL RANGKVITNK AVKGKYKFLQ KYYHRGAFFM DEDEEVYKRD FSAPTLEDHF NKTILPKVMQ VKNFGRSG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
MFAP1 is a component of the elastin-associated microfibrils. Microfibrils, found either in association with elastin or independently, are an important component of the extracellular matrix of many tissues. MFAP1 is one of a number of proteins demonstrated to be essential for cell division.
Specifications
| MFAP1 | |
| Polyclonal | |
| Unconjugated | |
| MFAP1 | |
| 4432409M24Rik; Mfap1; Mfap1a; Mfap1b; microfibril associated protein 1; microfibrillar associated protein 1; microfibrillar-associated protein 1; microfibrillar-associated protein 1A; Microfibrillar-associated protein 1B; microfibrillar-associated protein 1-like protein; RGD1562232; Spliceosome B complex protein MFAP1; Spliceosome B complex protein MFAP1A; Spliceosome B complex protein MFAP1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55081 | |
| MFAP1 | |
| Recombinant protein fragment of Human MFAP1 (Product #RP-90794) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction