Learn More
MFAP3 Antibody (OTI1B3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72657AF405
Product Code. 18129015
  • € 751.00 / 0.10mL
Description
MFAP3 Monoclonal specifically detects MFAP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MFAP3
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
MFAP3
Full length recombinant protein of human MFAP3 (NP_005918) produced in HEK293T cells.
0.1 mL
4238.0
Store at 4°C in the dark.
Western Blot, Immunocytochemistry, Immunofluorescence
OTI1B3
Western Blot, Immunocytochemistry/Immunofluorescence
DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3
Mouse
Affinity Purified