missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP3 Antibody (OTI1B3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72657PCP
This item is not returnable.
View return policy
Description
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MFAP3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4238 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI1B3 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
| Full length human recombinant protein of human MFAP3(NP_005918) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction