missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP3 Monoclonal Antibody (OTI1B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506942S
This item is not returnable.
View return policy
Description
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| MFAP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BR39 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI1B3 | |
| Unconjugated | |
| JPH2 | |
| MFAP3; Microfibril-associated glycoprotein 3; microfibrillar associated protein 3; microfibrillar-associated protein 3 | |
| Full length human recombinant protein of human JPH2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction