missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP3L Antibody (1059143), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB113251Y
This item is not returnable.
View return policy
Description
MFAP3L Monoclonal antibody specifically detects MFAP3L in Human samples. It is validated for Flow Cytometry
Specifications
| MFAP3L | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9848 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 1059143 | |
| Flow Cytometry | |
| HSD39, HSD-39, microfibrillar-associated protein 3-like, NYD-sp9, testis development protein NYD-SP9 | |
| Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction