missing translation for 'onlineSavingsMsg'
Learn More

MFAP4 Antibody (12D11), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42638AF532

Product Code. 18114768

  • € 592.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MFAP4
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
MFAP4
Recombinant human MFAP4 (22-255aa) purified from E. coli
0.1 mL
Primary
Human
IgG1 κ
Western Blot, ELISA
12D11
Western Blot, ELISA
microfibril-associated glycoprotein 4, microfibrillar-associated protein 4
Mouse
Protein A purified
Signal Transduction
4239.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.