missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP4 Antibody (12D11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42638FR
This item is not returnable.
View return policy
Description
MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MFAP4 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MFAP4 | |
| Recombinant human MFAP4 (22-255aa) purified from E. coli | |
| 100 μL | |
| Signal Transduction | |
| 4239 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 12D11 | |
| Western Blot, ELISA | |
| microfibril-associated glycoprotein 4, microfibrillar-associated protein 4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction